Anti-TSC-1 Rabbit Monoclonal Antibody [clone: EPR19260-105]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR19260-105] to TSC-1 - BSA and Azide free (Detector).
Type: Primary
Antigen: TSC-1
Clonality: Monoclonal
Clone: EPR19260-105
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Monkey
Frequently Bought Together