Anti-VCL Rabbit Monoclonal Antibody [clone: EPR20407]
: Abcam
New Product
Rabbit monoclonal [EPR20407] to Vinculin.
Type: Primary
Antigen: VCL
Clonality: Monoclonal
Clone: EPR20407
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together