Order Entry
Poland
ContactUsLinkComponent
Anti-IGHG4 Rabbit Monoclonal Antibody [clone: MOPC-21]
Anti-IGHG4 Rabbit Monoclonal Antibody [clone: MOPC-21]
Catalog # ABCAAB242443-200
Supplier:  Abcam
undefined
Anti-IGHG4 Rabbit Monoclonal Antibody [clone: MOPC-21]
Catalog # ABCAAB242443-200
Supplier:  Abcam

Specifications

  • Size:
    200 µg
  • Pack type:
    Vial

Specifications

About this item

Rabbit monoclonal, [MOPC-21] Isotype Control.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: IGHG4
Clonality: Monoclonal
Clone: MOPC-21
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: