Order Entry
Poland
ContactUsLinkComponent
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Catalog # ABCAAB18443-500
Supplier:  Abcam
undefined
Anti-IgG1 Mouse Monoclonal Antibody [clone: MOPC-21]
Catalog # ABCAAB18443-500
Supplier:  Abcam

Specifications

  • Seq:
    1418
  • Size:
    500 µg
  • Pack type:
    Vial

Specifications

About this item

Mouse IgG1, kappa monoclonal [MOPC-21] - isotype control.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: