Order Entry
Poland
ContactUsLinkComponent
Anti-Kallikrein 14 Rabbit Polyclonal Antibody
Anti-Kallikrein 14 Rabbit Polyclonal Antibody
Catalog # ABCAAB232966-100
Supplier:  Abcam
undefined
Anti-Kallikrein 14 Rabbit Polyclonal Antibody
Catalog # ABCAAB232966-100
Supplier:  Abcam

Specifications

  • Size:
    100 µg
  • Pack type:
    Vial

Specifications

About this item

Rabbit polyclonal to Kallikrein 14.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Kallikrein 14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Pig