Recombinant Human DLL3 (from HEK293 cells)
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Ice Buckets, Rectangular, with Lid
Supplier: VWR Collection
Centrifuge tubes
Supplier: Brand
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
VWR®, Cryo Boxes
Supplier: VWR Collection
Microplates, 96-well
Supplier: RATIOLAB
VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection