Human Recombinant Autotaxin (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human Autotaxin, expressed in HEK293 cells, suitable for WB, SDS-PAGE, Immunoassays, Functional Studies and is biologically active.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Upon delivery aliquot and store at –20 or –70 °C. Avoid freeze/thaw cycles.
- Endotoxin content:<0,1 ng/µg (1 IEU/µg), as measured by LAL method.
- Gene ID:UniprotID# Q13822
- Carrier-free:N
- Protein synonyms:ATX|Autotaxin|E-NPP 2|Ectonucleotide pyrophosphatase/phosphodiesterase family member 2|Extracellular lysophospholipase D|LysoPLD|PDNP2
- Protein/peptide name:Autotaxin
- Purity:>95% (by SDS-PAGE)
- Molecular weight:96 kDa (including tags)
- Sequence:DSPWTNISGSCKGRCFELQEAGPPDCRCDNLCKSYTSCCHDFDELCLKTARGWECTKDRCGEVRNEENACHCSEDCLARGDCCTNYQVVCKGESHWVDDDCEEIKAAECPAGFVRPPLIIFSVDGFRASYMKKGSKVMPNIEKLRSCGTHSPYMRPVYPTKTFPNLYTLATGLYPESHGIVGNSMYDPVFDATFHLRGREKFNHRWWGGQPLWITATKQGVKAGTFFWSVVIPHERRILTILQWLTLPDHERPSVYAFYSEQPDFSGHKYGPFGPEMTNPLREIDKIVGQLMDGLKQLKLHRCVNVIFVGDHGMEDVTCDRTEFLSNYLTNVDDITLVPGTLGRIRSKFSNNAKYDPKAIIANLTCKKPDQHFKPYLKQHLPKRLHYANNRRIEDIHLLVERRWHVARKPLDVYKKPSGKCFFQGDHGFDNKVNSMQTVFVGYGSTFKYKTKVPPFENIELYNVMCDLLGLKPAPNNGTHGSLNHLLRTNTFRPTMPEEVTRPNYPGIMYLQSDFDLGCTCDDKVEPKNKLDELNKRLHTKGSTEERHLLYGRPAVLYRTRYDILYHTDFESGYSEIFLMPLWTSYTVSKQAEVSSVPDHLTSCVRPDVRVSPSFSQNCLAYKNDKQMSYGFLFPPYLSSSPEAKYDAFLVTNMVPMYPAFKRVWNYFQRVLVKKYASERNGVNVISGPIFDYDYDGLHDTEDKIKQYVEGSSIPVPTHYYSIITSCLDFTQPADKCDGPLSVSSFILPHRPDNEESCNSSEDESKWVEELMKMHTARVRDIEHLTSLDFFRKTSRSYPEILTLKTYLHTYESEIGSGHHHHHHHHHH
- Concentration:Lot specific
- Formulation:Supplied in Phosphate Buffered Saline with 20% Glycerol.
- Shipping temperature:Shipped on blue ice at 4 °C.
- Tested applications:WB, SDS-PAGE, Immunoassays, Functional Studies
Specifications
Frequently Bought Together