Human Synthetic PAR2 (from HEK293 Cells)
New Product
Full length human PAR2, expressed in HEK293 cells, suitable for Western Blot, ELISA.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P55085
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Coagulation factor II receptor-like 1|F2RL1|G-protein coupled receptor 11|GPR11|PAR-2|Proteinase-activated receptor 2|Thrombin receptor-like 1
- Protein/peptide name:PAR2
- Molecular weight:Full length human PAR2 protein has a MW of 44,1 kDa.
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:Western Blot, ELISA
Frequently Bought Together