Recombinant Human CD30 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
VWR®, Cryo Boxes
Supplier: VWR Collection
for Portioncup 723062/201 1 * 2.500 items
Supplier: KING DISPOSABLES
Cellulose stoppers, Steristoppers®
Supplier: HERENZ HEINZ
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific
Seals for ALPS™ 50 V heat sealer
Supplier: THERMO MATRIX TECHNOLOGIES
Microplates, 96-well with glass inserts
Supplier: HIRSCHMANN
Cellulose chromatography papers, Whatman™
Supplier: Whatman products (Cytiva)
Screw caps for T501 sample tubes
Supplier: Simport Scientific
Test tubes with screw caps
Supplier: BOETTGER PAUL
CapInsert™ cap inserts for Cryovial® tubes
Supplier: Simport Scientific
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Sample container with snap-on cap
Supplier: VITLAB
Cryoboxes
Supplier: RATIOLAB
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning