Recombinant Human CDCP1 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
Cell culture plates, 96-well, CELLSTAR®
Supplier: Greiner Bio-One
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Polystyrene Assay Plates, 96- and 384-Well
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Lab blender bags, Stomacher®
Supplier: Seward (FermionX)
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, Cell Strainers
Supplier: VWR Collection
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Sample container with snap-on cap
Supplier: VITLAB
Accessories and consumables for KingFisher™ Flex
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection