Human Synthetic TA2R3 (from HEK293 Cells)
New Product
Full length human TA2R3, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9NYW6
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein/peptide name:TA2R3
- Molecular weight:Full length human TA2R3 protein has a MW of 35,9 kDa.
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
Frequently Bought Together