Order Entry
Poland
ContactUsLinkComponent
 
Native Botulinum C3 Enzyme (from Bacteria)
  ANTIA121191-10
New Product
 :  
Native Botulinum C3 Enzyme (from Bacteria)
  ANTIA121191-10
New Product
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Native
  • Source:
    Bacteria
  • Storage conditions:
    Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P15879
  • Carrier-free:
    N
  • Protein/peptide name:
    Botulinum C3 Enzyme
  • Purity:
    >90% (by SDS-PAGE)
  • Molecular weight:
    24 kDa
  • Concentration:
    1,33 mg/ml
  • Formulation:
    Supplied in 5 mM Sodium phosphate buffer, pH 6,0, with 50% Glycerol
  • Shipping temperature:
    Shipped on blue ice at 4 °C.
  • Tested applications:
    SDS-PAGE, Functional Studies

 

 

Full length Botulinum C3 Enzyme suitable for SDS-PAGE, Functional Studies and is biologically active.

 : This product is for research use only. It is not for use in diagnostic or therapeutic procedures.