Order Entry
Poland
ContactUsLinkComponent
 
Cynomolgus macaque Recombinant Eotaxin 2 (from HEK293 cells)
New Product
 
Cynomolgus macaque Recombinant Eotaxin 2 (from HEK293 cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus macaque
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# O00175
  • Reconstitution instructions:
    Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
  • Carrier-free:
    N
  • Protein synonyms:
    C-C motif chemokine 24|CCL24|CK-beta-6|Eosinophil chemotactic protein 2|Eotaxin-2|MPIF-2|MPIF2|Myeloid progenitor inhibitory factor 2|SCYA24|Small-inducible cytokine A24
  • Protein/peptide name:
    Eotaxin 2
  • Purity:
    >95% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 36,6 kDa after removal of the signal peptide. The apparent molecular mass of cCCL24-hFc is approximately 35 - 55 kDa due to glycosylation.
  • Sequence:
    CCL24 (Val27-His Tag119) + hFc (Glu99-Ala330)
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    SDS-PAGE

 

Frequently Bought Together