Human Recombinant Clusterin (from HEK293 Cells)
New Product
Partial length Human Clusterin, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P10909
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:Aging-associated gene 4 protein|Apo-J|APOJ|Apolipoprotein J|CLI|CLU|Complement cytolysis inhibitor|Complement-associated protein SP-40,40|Ku70-binding protein 1|KUB1
- Protein/peptide name:Clusterin
- Purity:>95% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 50,9 kDa after removal of the signal peptide. The apparent molecular mass of CLU-His is approximately 35 - 55 kDa due to glycosylation.
- Sequence:CLU (Asp23-Glu449) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Frequently Bought Together