Human Recombinant FGFR2b (from HEK293 Cells)
Dostawca: Antibodies.com
New Product
Certyfikaty
Informacje o tym produkcie
Partial length Human FGFR2b, expressed in HEK293 cells, suitable for WB, SDS-PAGE, immunoassays, functional studies and is biologically active.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Dane techniczne
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Upon delivery aliquot and store at −20...−70 °C. Avoid freeze/thaw cycles.
- Endotoxin content:<0,1 ng/µg (1 IEU/µg) as measured by LAL method
- Gene ID:UniprotID# P21802-3
- Carrier-free:N
- Protein/peptide name:FGFR2b
- Purity:>95% (by SDS-PAGE)
- Molecular weight:41,2 kDa (including tags)
- Sequence:RPSFSLVEDTTLEPEEPPTKYQISQPEVYVAAPGESLEVRCLLKDAAVISWTKDGVHLGPNNRTVLIGEYLQIKGATPRDSGLYACTASRTVDSETWYFMVNVTDAISSGDDEDDTDGAEDFVSENSNNKRAPYWTNTEKMEKRLHAVPAANTVKFRCPAGGNPMPTMRWLKNGKEFKQEHRIGGYKVRNQHWSLIMESVVPSDKGNYTCVVENEYGSINHTYHLDVVERSPHRPILQAGLPANASTVVGGDVEFVCKVYSDAQPHIQWIKHVEKNGSKYGPDGLPYLKVLKHSGINSSNAEVLALFNVTEADAGEYICKVSNYIGQANQSAWLTVLPKQQAPGREKEITASPDYLEGSGHHHHHHHHHH
- Concentration:Lot specific
- Formulation:Supplied in phosphate buffered saline with 20% glycerol
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:WB, SDS-PAGE, Immunoassays, Functional Studies
Dane techniczne
Frequently Bought Together