Human Synthetic Tspan-8 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Full length Human Tspan-8, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P19075
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:Tetraspanin-8|TM4SF3|Transmembrane 4 superfamily member 3|Tumor-associated antigen CO-029
- Protein/peptide name:Tspan-8
- Molecular weight:Full length human Tspan-8 protein has a MW of 26,1 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilization buffer (20 mM tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together