Human Recombinant GAS 6 (from HEK293 Cells)
New Product
Partial length human GAS 6, expressed in HEK293 cells, suitable for SDS-PAGE, Flow Cytometry.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q14393
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:GAS 6
- Protein/peptide name:GAS 6
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 72,7 kDa after removal of the signal peptide. The apparent molecular mass of GAS6-His is approximately 70 - 100 kDa due to glycosylation.
- Sequence:GAS6 (Ala31-Ala678) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE, Flow Cytometry
Frequently Bought Together