Human Synthetic OR11A1 (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Full length Human OR11A1, expressed in HEK293 cells.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q9GZK7
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:Hs6M1-18|Olfactory receptor 11A1|Olfactory receptor 11A2|Olfactory receptor OR6-30|OR11A2
- Protein/peptide name:OR11A1
- Molecular weight:Full length human OR11A1 protein has a MW of 35,3 kDa.
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C.
Specifications
Frequently Bought Together