Order Entry
Poland
ContactUsLinkComponent
Human Recombinant FGFR4 (from HEK293 cells)
Human Recombinant FGFR4 (from HEK293 cells)
Supplier:  Antibodies.com
New Product
Certificates
Human Recombinant FGFR4 (from HEK293 cells)
Supplier:  Antibodies.com
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Upon delivery aliquot and store at −20 or −70 °C. Avoid freeze/thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg), as measured by LAL method
  • Gene ID:
    UniprotID# P22455
  • Carrier-free:
    N
  • Protein synonyms:
    CD334|FGFR-4|Fibroblast growth factor receptor 4|JTK2|TKF
  • Protein/peptide name:
    FGFR4
  • Purity:
    >95% (by SDS-PAGE)
  • Molecular weight:
    40,1 kDa (Including tags)
  • Sequence:
    LEASEEVELEPCLAPSLEQQEQELTVALGQPVRLCCGRAERGGHWYKEGSRLAPAGRVRGWRGRLEIASFLPEDAGRYLCLARGSMIVLQNLTLITGDSLTSSNDDEDPKSHRDLSNRHSYPQQAPYWTHPQRMEKKLHAVPAGNTVKFRCPAAGNPTPTIRWLKDGQAFHGENRIGGIRLRHQHWSLVMESVVPSDRGTYTCLVENAVGSIRYNYLLDVLERSPHRPILQAGLPANTTAVVGSDVELLCKVYSDAQPHIQWLKHIVINGSSFGADGFPYVQVLKTADINSSEVEVLYLRNVSAEDAGEYTCLAGNSIGLSYQSAWLTVLPEEDPTWTAAAPEARYTDGSGHHHHHHHHHH
  • Concentration:
    Lot specific
  • Formulation:
    Supplied in phosphate buffered saline with 20% Glycerol
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    WB, SDS-PAGE, Immunoassays, Functional Studies

Specifications

Frequently Bought Together