Human Synthetic TPRA1 (from HEK293 Cells)
New Product
Full length Human TPRA1, expressed in HEK293 cells.
: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q86W33
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:GPR175|Integral membrane protein GPR175|TMEM227|TPRA1|Transmembrane protein 227|Transmembrane protein adipocyte-associated 1
- Protein/peptide name:TPRA1
- Molecular weight:Full length human TPRA1 protein has a MW of 41,1 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilization buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8.0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
Frequently Bought Together