Plants Recombinant Phl p 12 (12.0101) (from E. coli)
ANTIA254407-100
New Product
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Plants
- Seq:1286
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Use immediately or aliquot and store at −20...−80 °C. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P35079
- Reconstitution instructions:Reconstitute with sterile deionized water; With or without Sodium Azide
- Carrier-free:N
- Protein/peptide name:Phl p 12 (12.0101)
- Molecular weight:15,6 kDa
- Sequence:DNA sequence encoding 143 amino acids was fused with Strep Tag at the N-terminus to form this recombinant protein.
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from 100 mM Tris Buffer, pH 8, with 150 mM NaCl, and without preservatives or carriers (0,2 µm filter sterilised)
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA
Phleum pratense (Timothy Grass) Phl p 12 (12.0101), expressed in Escherichia coli, suitable for ELISA.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.