Order Entry
Poland
ContactUsLinkComponent
 
Human Synthetic CD39 (from HEK293 cells)
New Product
 
Human Synthetic CD39 (from HEK293 cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# P49961
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    ATP diphosphohydrolase|ATP-DPH|ATPDase|Ecto-apyrase|Ecto-ATP diphosphohydrolase 1|Ecto-ATPase 1|Ecto-ATPDase 1|Ectonucleoside triphosphate diphosphohydrolase 1|ENTPD1|Lymphoid cell activation antigen
  • Protein/peptide name:
    CD39
  • Molecular weight:
    Full length human CD39 protein has a MW of 58 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

 

Frequently Bought Together