Human Synthetic MLC1 (from HEK293 Cells)
New Product
Full length Human MLC1, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# Q15049
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:KIAA0027|Megalencephalic leukoencephalopathy with subcortical cysts protein 1|Membrane protein MLC1|WKL1
- Protein/peptide name:MLC1
- Molecular weight:Full length human MLC1 protein has a MW of 41,2 kDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from nanodisc solubilization buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA, SDS-PAGE
Frequently Bought Together