Synthetic Human LPA-4 (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
Serological pipettes, Nunc™
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Microspatulas, LaboPlast® / SteriPlast®
Supplier: BURKLE
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
Supplier: Corning
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Lids for 96-well microplates
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Sample tubes
Supplier: Simport Scientific