Order Entry
Poland
ContactUsLinkComponent
 
Human Synthetic BAI3 (from HEK293 cells)
New Product
 
Human Synthetic BAI3 (from HEK293 cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Synthetic
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# O60242
  • Reconstitution instructions:
    Reconstitute with distilled sterile water
  • Carrier-free:
    N
  • Protein synonyms:
    ADGRB3|Adhesion G protein-coupled receptor B3|Brain-specific angiogenesis inhibitor 3|KIAA0550
  • Protein/peptide name:
    BAI3
  • Molecular weight:
    Full length human BAI3 protein has a MW of 171,5 kDa
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilised from nanodisc solubilisation buffer (20 mM Tris-HCl, 150 mM NaCl, pH 8,0). Normally 5 - 8% trehalose is added as protectants before lyophilisation.
  • Shipping temperature:
    Shipped on blue ice at 4 °C

 

Frequently Bought Together