Synthetic Human CRLR (from HEK293 cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Frequently Bought Together
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Petri dishes, Corning® Gosselin™
Supplier: Corning
Cell scrapers
Supplier: Biotium
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
VIAL 55ML CUSTODY CLEAR 19 1 * 450 items
Supplier: Capitol Europe
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific