Order Entry
Poland
ContactUsLinkComponent
Adapter, delivery, bent, Quickfit®
Adapter, delivery, bent, Quickfit®
Catalog # BIBBRA12/2
Dostawca:  SciLabware
undefined
Adapter, delivery, bent, Quickfit®
Catalog # BIBBRA12/2
Dostawca:  SciLabware
Dane techniczne
  • Cone size:
    19/26 NS
  • Socket size:
    19/26 NS
  • Pk:
    5
Dane techniczne
Informacje o tym produkcie
PYREX®, borosilicate glass, bend.
  • Excellent chemical resistance
  • High temperature resistance
Receiver RA12/2 is designed to accompany the vacuum receiver (RA11/SC) for four flasks shown in the image. It has an extension below the cone which is bent to direct the flow of condensate to the individual outlet chosen.