Order Entry
Poland
ContactUsLinkComponent
Condensers, cold finger
Condensers, cold finger
Catalog # 201-8389
Supplier:  SciLabware
undefined
Condensers, cold finger
Catalog # 201-8389
Supplier:  SciLabware
Specifications
  • Cone size:
    19/26 NS
  • Jacket length:
    100 mm
  • Pk:
    1
Specifications
About this item
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it.