Filtering apparatus, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
Filtration apparatus with unit made from clear DURAN® borosilicate glass 3.3, PTFE insert and anodised aluminium clamp.
- Scaled funnel
- All components are available as spare parts
Typical applications: Coarse and ultrafine filtration, filtration of HPLC media, residue analysis. Filter paper, membrane filters (47 mm) or glass filters can be used for filtration.
Frequently Bought Together