Lithiumbis(trimethylsilyl)amide 1 mol/l in hexane, Sigma-Aldrich®
Catalog # SIAL224367-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:167.33 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:223-725-6
UN:2924
CAS Number:4039-32-1
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261