Order Entry
Poland
ContactUsLinkComponent
Lithiumbis(trimethylsilyl)amide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Lithiumbis(trimethylsilyl)amide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Catalog # SIAL577014-100ML
Supplier:  Merck
CAS Number:  
Lithiumbis(trimethylsilyl)amide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Catalog # SIAL577014-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :167.33 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:223-725-6
CAS Number:4039-32-1
Formula:C₆H₁₈LiNSi₂
MDL Number:MFCD00008261