Order Entry
Poland
ContactUsLinkComponent
tert-Butyldimethylsilyl trifluoromethanesulphonate
tert-Butyldimethylsilyl trifluoromethanesulphonate
Catalog # MOLEM84017698
Supplier:  Molekula
CAS Number:  
tert-Butyldimethylsilyl trifluoromethanesulphonate
Catalog # MOLEM84017698
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :264.34 g/mol
EINECS:274-102-0
UN:2920
ADR:8,II
CAS Number:69739-34-0
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000405