Glycinamide hydrochloride (H-Gly-NH2.HCl), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1668-10-6
:
Formula:C₂H₇ClN₂O
MDL Number:MFCD00013008
:110.54 g/mol
:M-Clarity™ quality level = MQ100
EINECS:216-789-1
Frequently Bought Together