Order Entry
Poland
ContactUsLinkComponent
 
Calcium sulphate, Drierite™ 10-20 mesh, without indicator
 
undefined
Calcium sulphate, Drierite™ 10-20 mesh, without indicator

 

  • Appearance:
    White to off-white Irregular granules
  • Water:
    ≤0.3 %
  • Particle size:
    10 to 20 mesh
  • Composition:
    ≥99.5 % (CaSO4) Typical

 

 

Frequently Bought Together