Order Entry
Poland
Orders LinkContactUsLinkComponent
Top tilt measures
Top tilt measures
Top tilt measures
Certificates
Top tilt measures
Top tilt measures
6 Options Available Below
Product Details & Documents
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 5 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 140 zł
    140 zł
    1 * 1 SZT
       140 zł
  • 10 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 145 zł
    145 zł
    1 * 1 SZT
       145 zł
  • 20 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 160 zł
    160 zł
    1 * 1 SZT
       160 zł
  • 25 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 165 zł
    165 zł
    1 * 1 SZT
       165 zł
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 197 zł
    197 zł
    1 * 1 SZT
       197 zł
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 371 zł
    371 zł
    1 * 1 SZT
       371 zł