Storage tubes, MATRIX™
About this item
PP, translucent
- Non sterile or sterile available
- Tubes are 100% leak tested to ensure sample integrity and security
- Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or deep well block sample storage.
14 Options Available Below
Product Details & Documents
PP, translucent
- Non sterile or sterile available
- Tubes are 100% leak tested to ensure sample integrity and security
- Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or deep well block sample storage.
Specifications for Product Family
Specifications
- Catalog No:
- MATR4170
- MATR4271
- MATR4272
- MATR4273
- MATR4274
- MATR4140
- MATR4247
- MATR4248
- MATR4249
- MATR4250
- MATR4252
- MATR4147
- MATR4148
- MATR4253
- Environmentally Preferable:
- Low Manufacturing Impact|Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact|Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
- CapacityPackedTypeVersionDostępnośćPrice
- Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.MATR4170Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure - 0,75 ml10 latch racks of 96 tubes/caseBlank storage tubeNon sterileLog in for availability1 * 960 SZT1 200 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4271Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 0,75 ml10 latch racks of 96 tubes/casBlank storage tubeSterileLog in for availability1 * 960 SZT1 280 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4272Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 0,75 ml10 latch racks of 96 tubes/caseAlphanumeric storage tubeNon sterileLog in for availability1 * 960 SZT1 590 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4273Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 0,75 ml10 latch racks of 96 tubes/caseAlphanumeric storage tubeSterileLog in for availability1 * 960 SZT1 700 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4274Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.MATR4140Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationNAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure - 1,4 ml10 latch racks of 96 tubes/caseBlank storage tubeNon sterileLog in for availability1 * 960 SZT1 200 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4247Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 latch racks of 96 tubes/caseBlank storage tubeSterileLog in for availability1 * 960 SZT1 280 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4248Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 refill carriers of 96 tubesBlank storage tubeNon sterileLog in for availability1 * 960 SZT680 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4249Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 refill carriers of 96 tubesBlank storage tubeSterileLog in for availability1 * 960 SZT761 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4250Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 latch racks of 96 tubes/caseAlphanumeric storage tubeSterileLog in for availability1 * 960 SZT1 700 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4252Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 snap racks of 96 tubes/caseAlphanumeric storage tubeNon sterileLog in for availability1 * 960 SZT1 820 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4147Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 snap racks of 96 tubes/caseAlphanumeric storage tubeSterileLog in for availability1 * 960 SZT1 920 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4148Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 1,4 ml10 refill carriers of 96 tubesAlphanumeric storage tubeNon sterileLog in for availability1 * 960 SZT1 030 złSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR4253Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact