Order Entry
Poland
ContactUsLinkComponent
 
Sodium chlorite 25% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck
 
Sodium chlorite 25% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Boiling Point:
    100 °C
  • Density:
    1.21 g/cm3 (20 °C)

 

 

Frequently Bought Together