Sulphosuccinic acid ≥70% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5138-18-1
:
Formula:C₄H₆O₇S
MDL Number:MFCD00075139
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:225-898-3
UN:3265
Frequently Bought Together