Order Entry
Poland
ContactUsLinkComponent
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate, Sigma-Aldrich®
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate, Sigma-Aldrich®
Catalog # SIAL540129-5G
Supplier:  Merck
CAS Number:  
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate, Sigma-Aldrich®
Catalog # SIAL540129-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :228.26 g/mol
EINECS:417-210-7
CAS Number:70987-78-9
Formula:C₁₀H₁₂O₄S
MDL Number:MFCD00064489