Order Entry
Poland
ContactUsLinkComponent
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems | UV Transilluminators
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems
Catalog # 733-2310
undefined
UV transilluminators, slimline, single wavelength, for use with Syngene InGenius and U:Genius systems
Catalog # 733-2310
Specifications
  • Model:
    GE-2020M-E
  • Wavelength:
    302 nm
  • Description:
    UV transilluminator, with runners
  • Seq:
    01
  • Filter size:
    200×200 mm
  • Pk:
    1