Order Entry
Poland
ContactUsLinkComponent
1-Dodecene-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
1-Dodecene-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL491209-100MG
Supplier:  Merck
CAS Number:  
1-Dodecene-[1,2-¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL491209-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:198332-88-6
Formula:C₁₂H₂₄