(R)-(+)-3,3'-Dibromo-1,1'-bi-2-naphthol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111795-43-8
:
Formula:C₂₀H₁₂Br₂O₂
MDL Number:MFCD03093629
:444.12 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together