1-Amino-4-naphthol hydrochloride, Sigma-Aldrich®
Catalog # SIAL133485-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:195.65 g/mol
EINECS:227-727-8
CAS Number:5959-56-8
Formula:C₁₀H₁₀ClNO
MDL Number:MFCD00012562