
4-(4-Nitrophenylazo)-1-naphthol for synthesis, Sigma-Aldrich®
Catalog # 8.41435.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Melting Point:277 - 280 °C (decomposition)
- Molar Mass:293.28 g/mol
Specification Test Results
About this item
:
:293.28 g/mol
:M-Clarity™ quality level = MQ200
EINECS:226-129-4
:GHS07
Storage Temperature:Ambient
CAS Number:5290-62-0
Formula:C₁₆H₁₁N₃O₃
MDL Number:MFCD00003975