
Diethyl (2-oxopropyl)phosphonate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1067-71-6
:
Formula:C₇H₁₅O₄P
MDL Number:MFCD00044728
:194.17 g/mol
Boiling Pt:98 °C (1 mmHg)
Density:1.11 g/cm³
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4325 to 1.4355 (20°C, 589 nm)
- GC:≥95 %
- Iron (Fe):≤0.0001 %
- Heavy metals (as Pb):≤1 ppm
Specification Test Results
Frequently Bought Together