4-(Aminomethyl)piperidine, Sigma-Aldrich®
Catalog # SIALA65158-5G-A
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.1888 g/mol
:M-Clarity™ quality level = MQ100
EINECS:230-446-3
CAS Number:7144-05-0
Formula:C₆H₁₄N₂
MDL Number:MFCD00006007