
Sodium deoxycholate, pure
Catalog # POCH796500428-100G
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:414.56 g/mol
Melting Pt:357…365 °C
EINECS:206-132-7
:GHS07
Storage Temperature:Ambient
CAS Number:302-95-4
Formula:C₂₄H₃₉NaO₄
MDL Number:MFCD00064139