
Lactic acid 20% in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:50-21-5
:598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520
:90.08 g/mol
EINECS:200-018-0
:GHS05
Storage Temperature:Ambient
Frequently Bought Together