Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
Frequently Bought Together