
Potassium thiocyanate, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1.61 g/cm³ (20 °C)
:97.18 g/mol
EINECS:206-370-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together